missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye 581-C3.5 NHS Ester
Click to view available options
:
Quantity: 1mg
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C61H90N6O16S4
- Mol. Weight: 1291.66g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 581nm
- Emission (Max): 596nm
- Solubility: DMSO, low in water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Specifications
| Description | Cyanine 3.5 dyes have enhanced fluorescence upon binding to proteins. C3.5 NHS Ester reacts with amino groups. |
| For Use With (Application) | Conjugation; in vivo Imaging |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 581-C3.5 NHS Ester |
| Molecular Weight (g/mol) | 1291.66g/mol |
| Purity | ≥90% |
| Content And Storage | Store in the dark at -20°C |
| Label or Dye | Fluorescent Dye 581-C3.5 NHS Ester |
| Emission | 596 |
| Excitation | 581 |
For research use only (RUO).
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction