missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye 555-C3 Alkyne
Click to view available options
:
Quantity: 1mg
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C36H42F3N3O9S2
- Mol. Weight: 781.86g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 555nm
- Emission (Max): 565nm
- Solubility: DMSO, water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Specifications
| Description | Cyanine 3 dyes have enhanced fluorescence upon binding to proteins. C3 Alkyne reacts with azide groups. |
| For Use With (Application) | Conjugation; in vivo Imaging |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 555-C3 Alkyne |
| Molecular Weight (g/mol) | 781.86g/mol |
| Purity | ≥90% |
| Content And Storage | Store in the dark at -20°C |
| Label or Dye | Fluorescent Dye 555-C3 Alkyne |
| Emission | 565 |
| Excitation | 555 |
For research use only (RUO).
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction