missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Abnova™ Fluorescent Dye 514-I Succinimidyl Ester
Beskrivning
Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 514
- Purity: ≥90%
- Molar Extinction Co-efficient: >80,000 M-1cm-1
- Excitation (Max): 518nm
- Emission (Max): 542nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifikationer
Specifikationer
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 514-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 514 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 514-I Succinimidyl Ester |
| Emission | 542 |
| Visa mer |
For research use only (RUO).
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?