Learn More
Abnova™ Fluorescent Dye 488-I Succinimidyl Ester
Description
Fluorescent Dye 488-I Succinimidyl Ester conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescent Dye 488-I Succinimidyl Ester is not affected by pH between 4 to 10. Fluorescent Dye 488-I Succinimidyl Ester of single isomer has much higher purity, with better performance consistency from batch to batch. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 488, Cy2, DyLight 488, FITC
- Purity: ≥90%
- Molar Extinction Co-efficient: >70,000 M-1cm-1
- Excitation (Max): 491nm
- Emission (Max): 514nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Fluorescent Dye 488-I Succinimidyl Ester conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescent Dye 488-I Succinimidyl Ester is not affected by pH between 4-10. Compared to Alexa Fluor 488, Fluorescent Dye 488-I Succinimidyl Ester of single isomer has much hihger purity, bringing better performance consistancy from batch to batch. |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 488-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 488, Cy2, DyLight 488, FITC |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 488-I Succinimidyl Ester |
| Emission | 514 |
| Show More |
For research use only (RUO).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.