Learn More
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks
GREENER_CHOICE
Product Code. 11914794
Product Code. 11914794
missing translation for 'mfr': Thermo Scientific™ AB0977
Only null left
Add to basket
Add to basket
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks contain a lockable lid and silicone tube plugs for security during transport.
  • 96 individual 0.65 or 1.2mL tubes
  • Alphanumeric grid-referencing molded into the rack and lid
  • Temperature range -170°C to +120°C
  • Heavy-duty rack in a microplate format and in accordance with SBS footprint
  • Stackable with or without lid
  • Open bottom to allow full robotic access to micro-tubes—each tube is fully supported to withstand pressure applied by robotic systems
  • Sealing options include MULTISIP™ septum plugs, silicone tube plugs, storage plate cap strips
Ordering Alerts:
Supplied in packs of 10 racks, 96 tubes each
TRUSTED_SUSTAINABILITY
Specifications
Description 1.2mL Grid-Referenced Cluster Tube Rack