missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ABCG4 Recombinant Protein

Brand:  Abnova™ H00064137-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123433

  • 359.00€ / 10µg

Estimated Shipment: 16-09-2022
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH41091
Solution
64137
ABCG4 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
WHITE2
ABCG4
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
96.8
8
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABCG4
Human
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers