Learn More
Abcam Rabbit specific HRP/DAB Detection IHC Kit, 15 mL
Product Code. 32112225 Shop All Abcam Products
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112225 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32112225 Supplier Abcam Supplier No. ab6426115mL
 Restricted Item
Estimated Shipment: 25-11-2026
Antibody, suitable for IHC-P, IHC-Fr, with over 200 citations.
  • Applications: IHC-P, IHC-Fr
Specifications
Quantity 15 mL