missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Assay Type: Sandwich
- Detection Method: Colorimetric
- Sample Types: Plasma, Serum, Cell culture supernatant
- Species Reactivity: Human
- Sensitivity: 0.55 ng/mL
- Range: 0.55 - 150 ng/mL
- Results Type: Quantitative
- Storage: Store at -20°C
Specifications
Specifications
| Assay Range | 0.55 to 150 ng/mL |
| Assay Sensitivity | 0.55 ng/mL |
| Content And Storage | Store at -20°C |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA Kit |
| Sample Type | Plasma, Serum, Cell Culture Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene Alias | Calumenin, Crocalbin, IEF SSP 9302, CALU |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?