Learn More
Tocris Bioscience™ AA 147
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
AA 147 is an endoplasmic reticulum (ER) proteostasis regulator. Activates the ATF6 transcriptional program independently of ER stress and/or global UPR activation. Selectively reduces secretion of amyloidogenic transthyretin (TTR) from HepG2 cells. Reduces amyloidogenic immunoglobulin light chain (ALLC) secretion from ALMC-2 cells (EC50 = 1.1 μM).
Specifications
Specifications
| Chemical Name or Material | N-(2-Hydroxy-5-methylphenyl)benzenepropanamide |
| CAS | 393121-74-9 |
| Quantity | 5 mg |
| Molecular Formula | C16H17NO2 |
| Formula Weight | 255.31 |
| Purity | 98% |