Learn More
Leica Microsystems A60F Stereo Microscope
Product Code. 11980878
Change view
Click to view available options
Includes:
Flexible support arm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11980878 Flexible support arm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11980878 Supplier Leica Microsystems Supplier No. 10450311
 Restricted Item
Estimated Shipment: 04-01-2027
Specifications
Height (Metric) 35.6 cm
Includes Flexible support arm
Length (Metric) 17.4 cm
Dimensions (L x W x H) 17.4 x 119.0 x 35.6 cm
Product Type Stereo microscope