Learn More
Miele A 841/3 Injector nozzle Ø 6 x 210 mm (20 off)
Brand:  Miele 11113150
440.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
A 841/3 Injector nozzle Ø 6 x 210 mm (20 off)
Product Code. 17888151
  • 512.00€ / Each
Only null left
Estimated Shipment: 11-03-2026