Learn More
A 769662, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | A 769662 |
| CAS | 844499-71-4 |
| Molecular Formula | C20H12N2O3S |
| Quantity | 10 mg |
| Synonym | 4-Hydroxy-3-(2'-hydroxy-1,1'-biphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 6,7-Dihydro-4-hydroxy-3-(2'-hydroxy[1,1'-biphenyl]-4-yl)-6-oxo-thieno[2,3-b]pyridine-5-carbonitrile |
| InChI Formula | InChI=1S/C20H12N2O3S/c21-9-14-18(24)17-15(10-26-20(17)22-19(14)25)12-7-5-11(6-8-12)13-3-1-2-4-16(13)23/h1-8,10,23H,(H2,22,24,25) |
| SMILES | [Na+].CCC(=O)O[C@H](C(C)C)OP(CCCCc1ccccc1)(=O)CC(=O)N2C([C@@H](=O)[O-])C[C@@H](C3CCCCC3)C2 |
| IUPAC Name | 4-hydroxy-3-[4-(2-hydroxyphenyl)phenyl]-6-oxo-7H-thieno[2,3-b]pyridine-5-carbonitrile |
| Molecular Weight (g/mol) | 360.39 |
| Formula Weight | 360.0569 |