missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 10% v/v aq. soln., Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 042552.M4

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15451817

  • 350.00€ / 20 L
 Restricted Item
Estimated Shipment: 28-08-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7664-93-9
20L
H2SO4
UN2796
Miscible in water,acetic acid and alcohol.
OS(=O)(=O)O
98.072
CHEBI:26836
Sulfuric acid
10% v/v aq. soln.
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.