missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tacrine hydrochloride, Thermo Scientific™
A potent centrally acting anticholinesterase
Brand: Thermo Scientific Alfa Aesar J60070.03
| Quantity | 1g |
|---|
This item is not returnable.
View return policy
Description
SolubilitySoluble in aqueous buffers
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 1684-40-8 | |
| MFCD00150067 | |
| UN2811 | |
| tacrine hydrochloride, 1,2,3,4-tetrahydroacridin-9-amine hydrochloride, tacrine hcl, hydroaminacrine, tenakrin, 9-amino-1,2,3,4-tetrahydroacridine hydrochloride, tacrine.hcl, unii-4966rng0bu, 1,2,3,4-tetrahydro-9-acridinamine hydrochloride, ccris 7250 | |
| ZUFVXZVXEJHHBN-UHFFFAOYSA-N | |
| hydrogen 1,2,3,4-tetrahydroacridin-9-amine chloride | |
| 2723754 | |
| ≥98% |
| C13H15ClN2 | |
| 1g | |
| 14,9024 | |
| Soluble in aqueous buffers | |
| [H+].[Cl-].NC1=C2C=CC=CC2=NC2=C1CCCC2 | |
| 234.73 | |
| 234.73 (Anhydrous) | |
| Tacrine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction