Learn More
Brady™ Polyolefin PermaSleeve™ Wire Marker Sleeve
Specifications
Specifications
| Certifications/Compliance | SAE-DTL-23053/5 Class 1, SAE-AS-81531, MIL-STD–202, UL Recognized |
| Description | Sleeve |
| Color | White |
| Finish | Matte |
| Material | Polyolefin |
| Print Technology | Dot Matrix, Thermal Transfer |
| For Use With (Application) | Wire identification and insulation purposes |
| For Use With (Equipment) | BMP™71, TLS2200 |
| Quantity | 1000/Pk. |
| Size (Metric) | 44.8 x 21.6 (W x H) |
| Show More |