Learn More
Thermo Scientific™ 9mm Certified Clear Wide Opening Screw Thread Kits
Product Code. 15359464
Click to view available options
Description:
2mL; Clear ID Vial; With 9mm Preslit PTFE/Silicone Septa/Cap
Assembled Convenience Kit, Certified 9 mm Screw Clear I-D Vial 2 mL with bonded PTFE/Silicone Cap
Assembled Kit, Certified 9mm Wide Opening Screw Vial, 2mL
Certified Target DP 1.5mL Clear High Recovery Vials, 9mm Screw Thread
Certified Target DP 2mL Clear I-D Vial Kit, 9mm Screw Thread
Convenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL Reservoir
Convenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir
Convenience Kit, 9mm Wide OpeningScrew Vial, 2mL
Convenience Kit, 9mm Wide OpeningScrew Vial, 2mL, Silanized
Convenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
Convenience Kit, Certified 9mm Wide Opening Total Recovery Screw Vial, w/ 10μL Reservoir
Closure Color:
Black
Blue
Gray
Septum:
Blue PTFE/White Silicone
Bonded Clear PTFE/Clear Silicone
Bonded Red PTFE/White Silicone
Bonded White Silicone/Clear PTFE MS Certified
Ivory PTFE/Red Rubber
PTFE/Silica
Red PTFE/White Silicone
Marking Spot:
No
Yes
Unit Size:
Pack of 100
Product Code. 15359464
Brand: Thermo Scientific™ MSCERT500038
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ 9mm Wide Opening Screw Thread Certified Vial Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
Unassembled kits
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers
Assembled kits
  • 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Gray
Description Convenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir
Color Clear
Material Glass, PTFE, Silicone
Pre-slit Yes
Septum Bonded Clear PTFE/Clear Silicone
No. per Pack 100
Closure Type Screw Cap
Marking Spot No