missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 96-Well Treated Tissue Culture Plate
Click to view available options
Quantity:
1 Plate
50 Plates/Cs.
Unit Size:
1 set
Pack of 50
Description
- Tissue culture plates, 96-well, intended for culturing anchorage-dependent cells, including stromal cells
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 1 Plate |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction