missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 96-Well Treated Tissue Culture Plate

Product Code. 17128061
Encompass
Click to view available options
Quantity:
1 Plate
50 Plates/Cs.
Unit Size:
1 set
Pack of 50
This item is not returnable. View return policy

Product Code. 17128061

Brand: STEMCELL Technologies 27135

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

96-well culture plates pretested for the ability to support the growth of anchorage-dependent cells

  • Tissue culture plates, 96-well, intended for culturing anchorage-dependent cells, including stromal cells

Specifications

For Use With (Application) Cell Culture
Quantity 1 Plate
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.