Learn More
Greiner Bio-One 96-well Non Binding Microplates
Product Code. 15901661
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Unit Size
15901661 Round Bottom (U-well) Pack of 40
13219672 Flat Bottom (F-well) Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15901661 Supplier Greiner Bio-One Supplier No. 650901
  • ONLINE EXCLUSIVE PRICE
    291.00€
    267.89€ / Pack of 40
    Save 23.11€
    7% Off
Estimated Shipment: 06-01-2027
Features long-term surface stability without degradation or leaching.
  • Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
  • Long-term surface stability without degradation or leaching
  • Higher assay sensitivity with reduced background
  • Significantly increase assay sensitivity
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Product Type Non-binding Microplate
Material Polystyrene
Volume (Metric) Well 40 to 280 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-binding
Total Volume (Metric) Well 323 μL
Well Shape Round Bottom (U-well)
Max. RCF 1000 x g (swinging-bucket rotor)
No. per Case 40
Quantity 40/Cs.
Well Color Clear
Well Design Chimney