Learn More
Greiner Bio-One 96-well High Binding Standard ELISA Microplates
Product Code. p-7075398
Click to view available options
Well Shape:
Square, Round Bottom (U-well)
Sterility:
Non-sterile
Well Design:
Round
Unit Size:
Pack of 40
Product Code. 10227261
Brand: Greiner Bio-One 650061
Only null left
Add to basket
Add to basket
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9mm
  • Specifically tre
Service Plan
No
Certifications
DIN ISO 9001
Services
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Binding Type High Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon 600
Well Shape Square, Round Bottom (U-well)
Quantity 40/Cs.
Well Design Round