Learn More
Greiner Bio-One 96-well High Binding Standard ELISA Microplates
Click to view available options
Well Shape:
Square, Round Bottom (U-well)
Sterility:
Non-sterile
Well Design:
Round
Unit Size:
Pack of 40
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Specifically tre
Service Plan
No
Certifications
DIN ISO 9001
Services
No
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Binding Type | High Binding |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Microlon 600 |
| Well Shape | Square, Round Bottom (U-well) |
| Quantity | 40/Cs. |
| Show More |