Learn More
Greiner Bio-One 96-well CrystalQuick™ Plus Crystallisation Microplates
Product Code. p-7075382
Click to view available options
Volume (Metric) Well:
3.9 μL
4.1 μL
Height (Metric):
3.9 mm
14.4 mm
Unit Size:
Pack of 40
Pack of 80
Product Code. 10389343
Brand: Greiner Bio-One 609130
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Plus Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
  • Non-sterile
  • Polystyrene
  • Good optical properties
  • Hydrophobic surface
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, simultaneous experiments are possible with the sitting drop and hanging drop method
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Clear
Material Polystyrene
Volume (Metric) Well 4.1 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 40
Quantity 40/Cs.
Height (Metric) 14.4 mm