Learn More
Greiner Bio-One 96-well CrystalQuick™ Plus Crystallisation Microplates
Product Code. 10389343
Change view
Click to view available options
Volume (Metric) Well:
3.9 μL
4.1 μL
Surface Treatment:
Hydrophobic
Hydrophobic LBR
Unit Size:
Pack of 40
Pack of 80
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Well Surface Treatment unitSize
10389343 4.1 μL Hydrophobic Pack of 40
10083973 3.9 μL Hydrophobic Pack of 80
10502374 4.1 μL Hydrophobic LBR Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10389343 Supplier Greiner Bio-One Supplier No. 609130
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Plus Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
  • Non-sterile
  • Polystyrene
  • Good optical properties
  • Hydrophobic surface
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, simultaneous experiments are possible with the sitting drop and hanging drop method
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Clear
Material Polystyrene
Volume (Metric) Well 4.1 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Hydrophobic
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 40
Quantity 40/Cs.
Height (Metric) 14.4 mm