Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. p-9045029 Shop All Corning Products
Click to view available options
Packaging:
20/Pk., 100/Cs.
Individually wrapped, 48/Cs.
Unit Size:
Pack of 100
Pack of 48
Product Code. 10530753
Brand: Corning™ 3603
Edge
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid With Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Packaging Individually wrapped, 48/Cs.
    Color Black, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Attachment and growth of anchorage-dependent cells
    No. per Case 48/Cs.
    No. per Pack 1/Pk.
    Quantity 48/Cs.