Learn More
Uvex 95980 Insole For Uvex Naturform - Width 11
Product Code. 17239021 Shop All Uvex Products
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17239021
Brand: Uvex 9598040
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Comfortable climatic insole with antistatic properties for Uvex Performance (Naturform)
  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9598.0
  • Sizes: 38 to 48
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Uvex Naturform
Material Microvelour
Size 40
Disposable Reusable
Gender Unisex
Fit Standard width
Latex Free Yes
Waterproof No