missing translation for 'onlineSavingsMsg'
Learn More

9-Decenoic Acid, TRC

Product Code. 30240539
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240539 25 g 25g
30240476 100 g 100g
30240520 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240539 Supplier Toronto Research Chemicals Supplier No. TRCD22560025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14436-32-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
Synonym Caproleic Acid
IUPAC Name dec-9-enoic acid
SMILES O=C(O)CCCCCCCC=C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H18O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2H,1,3-9H2,(H,11,12)
Formula Weight 170.1307
Chemical Name or Material 9-Decenoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.