Learn More
9-cis-Retinol Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 9-cis Retinol Acetate |
| CAS | 29584-22-3 |
| Molecular Formula | C22 H32 O2 |
| Quantity | 1 mg |
| Synonym | Retinol','acetate','9-cis-, 9-cis-Retinol acetate, 9-cis-Retinyl acetate, 9-cis-Vitamin A acetate, 9-cis-Vitamin A1 acetate, Zuretinol acetate |
| InChI Formula | InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9-,18-14+ |
| SMILES | CC(=O)OC\C=C(/C)\C=C\C=C(\C)/C=C/C1=C(C)CCCC1(C)C |
| IUPAC Name | [(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate |
| Molecular Weight (g/mol) | 328.49 |
| Formula Weight | 328.2402 |