Learn More
STEMCELL Technologies 9-cis Retinoic Acid
Description
- 9-cis-Retinoic acid activates all isoforms of the retinoic acid receptor (RAR) (Ki = 0.5-27 nM) as well as retinoid X receptor (RXR) isoforms (Ki = 3.8-12 nM) (Umemiya et al.; Wong et al.) The ability of RAR to heterodimerize with RXR, and the latter to heterodimerize with other partners allows 9-cis-retinoic acid to evoke a wide range of effects (Dawson et al.; Kane)
Specifications
Specifications
| CAS | 5300-03-8 |
| Synonym | 9-cis-retinoic acid; Aliretinoin; NSC 659772; Panretin |
| Quantity | 5 mg |
| Molecular Formula | C20H28O2 |
| Purity | ≥95% |