Learn More
9-Chloro Triamcinolone Acetonide 21-Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 9-Chloro Triamcinolone Acetonide 21-Acetate |
| CAS | 10392-75-3 |
| Molecular Formula | C26 H33 Cl O7 |
| Quantity | 10 mg |
| Synonym | Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-chloro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-chloro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-acetate (7CI,8CI), 1H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv. |
| InChI Formula | InChI=1S/C26H33ClO7/c1-14(28)32-13-20(31)26-21(33-22(2,3)34-26)11-18-17-7-6-15-10-16(29)8-9-23(15,4)25(17,27)19(30)12-24(18,26)5/h8-10,17-19,21,30H,6-7,11-13H2,1-5H3/t17-,18-,19-,21+,23-,24-,25-,26+/m0/s1 |
| SMILES | CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(Cl)[C@@H](O)C[C@]23C |
| IUPAC Name | [2-[(1S,2S,4R,8S,9S,11S,12R,13S)-12-chloro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate |
| Molecular Weight (g/mol) | 492.989 |
| Formula Weight | 492.1915 |