missing translation for 'onlineSavingsMsg'
Learn More

9,11-Anhydro Fusidic Acid, TRC

Product Code. 30335994
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335994 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335994 Supplier Toronto Research Chemicals Supplier No. TRCA63950025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,11-Anhydrofusidic Acid
CAS 74048-41-2
Molecular Formula C31 H46 O5
Quantity 25 mg
Synonym 29-Nordammara-9(11),17(20),24-trien-21-oic acid, 16-(acetyloxy)-3-hydroxy-, (3α,4α,8α,13α,14β,16β,17Z)- (9CI), ent-(17Z)-16α-(Acetyloxy)-3β-hydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-9(11),17(20),24-trien-21-oic acid, δ9(11)-Anhydrofusidic acid, (3alpha,4alpha,8alpha,13alpha,14beta,16beta,17Z)-16-(Acetyloxy)-3-hydroxy-29-nordammara-9(11),17(20),24-trien-21-oic Acid, Fusidic Acid EP Impurity L
InChI Formula InChI=1S/C31H46O5/c1-18(2)9-8-10-21(28(34)35)27-23-11-12-26-29(5)15-14-24(33)19(3)22(29)13-16-30(26,6)31(23,7)17-25(27)36-20(4)32/h9,12,19,22-25,33H,8,10-11,13-17H2,1-7H3,(H,34,35)/b27-21-/t19-,22-,23-,24+,25-,29-,30-,31-/m0/s1
SMILES C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)C2=CC[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C
IUPAC Name (2Z)-2-[(3R,4S,5S,8R,10S,13R,14S,16S)-16-acetyloxy-3-hydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,12,13,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid
Molecular Weight (g/mol) 498.6939
Formula Weight 498.3345
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.