missing translation for 'onlineSavingsMsg'
Learn More

(±)-9,10-Dihydrojasmonic Acid, TRC

Product Code. 30369170
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369170 50 mg 50mg
30372558 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369170 Supplier Toronto Research Chemicals Supplier No. TRCD45426550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-9,10-Dihydrojasmonic Acid
CAS 3572-64-3
Molecular Formula C12H20O3
Quantity 50 mg
Synonym (±)-3-Oxo-2-pentyl-cyclopentane-1-acetic Acid, DJA, 3-Oxo-2-pentylcyclopentaneacetic Acid, (±)-9,10-Dihydrojasmonic Acid, 2-Pentyl-3-oxocyclopentyl-1-acetic Acid, 2-Pentyl-3-oxocyclopentylacetic Acid, 2-n-Pentyl-3-oxocyclopentylacetic Acid, 3-Carboxymethyl-2-pentylcyclopentanone, 3-Oxo-2-pentyl-cyclopentaneacetic Acid
InChI Formula InChI=1S/C12H20O3/c1-2-3-4-5-10-9(8-12(14)15)6-7-11(10)13/h9-10H,2-8H2,1H3,(H,14,15)
SMILES CCCCCC1C(CCC1=O)CC(=O)O
Molecular Weight (g/mol) 212.29
Formula Weight 212.1412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.