Learn More
Thermo Scientific™ 8 mm Autosampler Inserts
Product Code. p-6062318
Click to view available options
Volume (Metric):
175 μL
200 μL
250 μL
Material:
Glass
Polypropylene
Silanized Glass
Base Style:
Conical
Flat Bottom
Polyspring
Quantity:
100
500
1000
Unit Size:
Pack of 100
Pack of 1000
Pack of 500
Product Code. 10119084
Brand: Thermo Scientific™ C4012465
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 125μL to 200μL.
These clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes ranging from 125-200μL.
  • Pulled point inserts are an economical choice for non-critical applications
TRUSTED_SUSTAINABILITY
Specifications
Description 200µL MicroSert Insert
Diameter (Metric) Inner 5 mm
Color Clear
Diameter (Metric) Outer 5 mm
Material Glass
Volume (Metric) 250 μL
Style MicroSert, Flat bottom, Glass
Volume (Metric) Residual <12 μL
Base Style Flat Bottom
Volume (Metric) Total 250 μL
Height (Metric) 31 mm
Quantity 500
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Type Micro Insert
Volume (Metric) Usable 200 μL
For Use With (Equipment) 8 mm HPLC, GC Autosampler Vial