Learn More
Tocris Bioscience™ 8MDP
GREENER_CHOICE
Potent equilibrative nucleoside transporter (ENT) inhibitor
Brand:  Tocris Bioscience™ 4588/10
Encompass
Product Code. 15746206
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
8MDP is a potent equilibrative nucleoside transporter 1 (ENT1) inhibitor (IC50 = 0.43 nM). Inhibits hENT1 and hENT2 uptake of [H3]uridine in K562 cells and K15NTD cells respectively.
Specifications
Specifications
2,2′,2′′,2′′′-[[4,8-Bis(hexahydro-1(2H)-azocinyl)pyrimido[5,4-d]pyrimidine-2,6-diyl]dinitrilo]tetrakisethanol
29491-75-6
10 mg