Learn More
8-Methylnonanal, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Methylnonanal |
| CAS | 3085-26-5 |
| Molecular Formula | C10H20O |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C10H20O/c1-10(2)8-6-4-3-5-7-9-11/h9-10H,3-8H2,1-2H3 |
| SMILES | CC(C)CCCCCCC=O |
| IUPAC Name | 8-methylnonanal |
| Molecular Weight (g/mol) | 156.27 |
| Formula Weight | 156.1514 |