Learn More
8-Hydroxypenillic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Hydroxypenillic Acid |
| CAS | 3053-85-8 |
| Molecular Formula | C9 H12 N2 O5 S |
| Quantity | 2.5 mg |
| Synonym | Hexahydro-2,2-dimethyl-5-oxoimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid |
| InChI Formula | InChI=1S/C9H12N2O5S/c1-9(2)4(7(14)15)11-5(17-9)3(6(12)13)10-8(11)16/h3-5H,1-2H3,(H,10,16)(H,12,13)(H,14,15) |
| SMILES | CC1(C)SC2C(NC(=O)N2C1C(=O)O)C(=O)O |
| IUPAC Name | 2,2-dimethyl-5-oxo-3,6,7,7a-tetrahydroimidazo[5,1-b][1,3]thiazole-3,7-dicarboxylic acid |
| Molecular Weight (g/mol) | 260.267 |
| Formula Weight | 260.047 |