missing translation for 'onlineSavingsMsg'
Learn More

8-Chloroadenosine, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001511
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001511 10 mg 10mg
30001529 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001511 Supplier Tocris Bioscience Supplier No. 4436/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cytotoxic nucleoside analog; inhibits RNA synthesis

Nucleoside analog; metabolized in vivo to 8-Chloro-ATP. Incorporates into RNA during transcription and inhibits RNA synthesis. Exhibits cytotoxicity in MM.1S, RPMI-8226 and U266 cancer cell lines; induces G2/M cell cycle arrest and mitotic catastrophe in A549 and H1299 cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34408-14-5
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.687
InChI Key MHDPPLULTMGBSI-FTWQFJAYSA-N
Synonym 8-chloroade-nosine
PubChem CID 13009312
IUPAC Name (2R,3R,4R,5R)-2-(6-amino-8-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(C(=N1)N)N=C(N2C3C(C(C(O3)CO)O)O)Cl

Specifications

Quantity 10 mg
Formula Weight 301.69
Percent Purity >98%
Chemical Name or Material 8-Chloroadenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.