missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-1-octanol, 98%

Product Code. 11402050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402050 25 mL 25mL
11492040 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402050 Supplier Thermo Scientific Alfa Aesar Supplier No. H28417.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23144-52-7 | C8H17ClO | 164.673 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23144-52-7
Molecular Formula C8H17ClO
Molecular Weight (g/mol) 164.673
MDL Number MFCD00040005
InChI Key YDFAJMDFCCJZSI-UHFFFAOYSA-N
Synonym 8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
PubChem CID 90012
IUPAC Name 8-chlorooctan-1-ol
SMILES C(CCCCCl)CCCO

Specifications

Density 0.976
Boiling Point 129°C to 130°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.458
Quantity 25 mL
Formula Weight 164.67
Percent Purity 98%
Chemical Name or Material 8-Chloro-1-octanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.