Learn More
8-Carboxyoctanol, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Carboxyoctanol |
| CAS | 3788-56-5 |
| Molecular Formula | C9H18O3 |
| Quantity | 500 mg |
| Synonym | 9-Hydroxynonanoic Acid, 9-Hydroxypelargonic Acid |
| InChI Formula | InChI=1S/C9H18O3/c10-8-6-4-2-1-3-5-7-9(11)12/h10H,1-8H2,(H,11,12) |
| SMILES | OCCCCCCCCC(=O)O |
| IUPAC Name | 9-hydroxynonanoic acid |
| Molecular Weight (g/mol) | 174.24 |
| Formula Weight | 174.1256 |