Learn More
7a-Acetylthio-17a-ethoxymethyl Testosterone, TRC
Specifications
Specifications
| Chemical Name or Material | 7alpha-Acetylthio-17alpha-ethoxymethyl Testosterone |
| CAS | 3070439-36-7 |
| Molecular Formula | C24H36O4S |
| Quantity | 50 mg |
| Synonym | Spironolactone EP Impurity I |
| SMILES | O[C@]1(COCC)[C@]([C@@]2([H])CC1)(CC[C@@]([C@]3(CC4)C)([H])[C@@]2([H])[C@@H](CC3=CC4=O)SC(C)=O)C |
| Molecular Weight (g/mol) | 420.61 |