missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 72-well Microtest (Terasaki) Plate
GREENER_CHOICE

Product Code. 10670742
Change view
Click to view available options
Well Count:
72
Unit Size:
Pack of 270
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10670742 72 Pack of 270
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10670742 Supplier Greiner Bio-One Supplier No. 654180

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Suitable for all applications for serological determination of HLA antigens

Alphanumeric well coding
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 10 L
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Tissue Culture
Type Microtest Plate
Well Count 72
For Use With (Application) HLA Antigen Determination
Includes Lid
No. per Case 270
No. per Pack 10
Temperature Range (Metric) -20°C to +60°C
Length (Metric) 83.3 mm
Width (Metric) 58 mm
Height (Metric) 11 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.