missing translation for 'onlineSavingsMsg'
Learn More

7-Phenylheptanoic acid, 97%

Product Code. p-7044174
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11301797

Brand: Thermo Scientific Alfa Aesar L05009.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40228-90-8 | C13H18O2 | 206.285 g/mol

7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40228-90-8
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00037801
InChI Key ZVSXKFNTWOIGJI-UHFFFAOYSA-N
Synonym benzeneheptanoic acid, phenylheptanoic acid, 7-phenyl-heptanoic acid, acmc-20alwn, 7-phenylheptanoic acid, xxc
PubChem CID 520987
IUPAC Name 7-phenylheptanoic acid
SMILES C1=CC=C(C=C1)CCCCCCC(=O)O

Specifications

Melting Point ∼20°C
Density 1.009
Boiling Point 147°C to 150°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 1 g
Beilstein 1957081
Solubility Information Insoluble in water.
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material 7-Phenylheptanoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.