missing translation for 'onlineSavingsMsg'
Learn More

7-Oxabicyclo[2.2.1]heptane, 98%

Product Code. 11435207
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11435207

Brand: Thermo Scientific Alfa Aesar B22612.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 279-49-2 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 279-49-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00082342
InChI Key YPWFNLSXQIGJCK-UHFFFAOYSA-N
Synonym 1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
PubChem CID 9234
IUPAC Name 7-oxabicyclo[2.2.1]heptane
SMILES C1CC2CCC1O2

Specifications

Density 0.968
Boiling Point 118°C to 120°C (713 mmHg)
Quantity 25 g
Refractive Index 1.448
UN Number UN3271
Formula Weight 98.15
Percent Purity 98%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.