missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Methyl-d3-guanosine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 7-Methyl-d3-guanosine |
| Molecular Formula | C11 D3 H12 N5 O5 |
| Quantity | 0.01 g |
| Synonym | 7-Methyl-d3-guanosine, N7-(Methyl-d3)guanosine, 7-Methylguanosine-D3 (7-methyl-D3), 2-Amino-1,6-dihydro-7-(methyl-d3)-6-oxo-9-β-D-ribofuranosylpurine |
| InChI Formula | InChI=1 S/C11H15N5O5/c1-15-3-16(8-5(15)9(20)14-11(12)13-8)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19 H,2H2,1H3,(H2-,12,13,14,20)/t4-,6-,7-,10-/m1/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])[n+]1 cn([C@@H]2 O[C@H](CO)[C@@H](O)[C@H]2 O)c3nc(N)nc([O-])c13 |
| IUPAC Name | 2-amino-9-[(2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7-(trideuteriomethyl)purin-7-ium-6-olate |
| Molecular Weight (g/mol) | 300.2858 |
| Formula Weight | 300.1261 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction