Learn More
7-Methoxy-1-[[3-methoxy-4-(phenylmethoxy)phenyl]methyl]-6-(phenylmethoxy)isoquinoline, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Methoxy-1-((3-methoxy-4-(phenylmethoxy)phenyl)methyl)-6-(phenylmethoxy)isoquinoline |
| CAS | 29973-94-2 |
| Molecular Formula | C32H29NO4 |
| Quantity | 10 mg |
| Synonym | 6-(Benzyloxy)-1-[4-(benzyloxy)-3-methoxybenzyl]-7-methoxyisoquinoline, 1-(3'-Methoxy-4'-benzyloxybenzyl)-6-benzyloxy-7-methoxyisoquinoline |
| InChI Formula | InChI=1S/C32H29NO4/c1-34-30-18-25(13-14-29(30)36-21-23-9-5-3-6-10-23)17-28-27-20-31(35-2)32(19-26(27)15-16-33-28)37-22-24-11-7-4-8-12-24/h3-16,18-20H,17,21-22H2,1-2H3 |
| SMILES | COc1cc(Cc2nccc3cc(OCc4ccccc4)c(OC)cc23)ccc1OCc5ccccc5 |
| IUPAC Name | 7-methoxy-1-[(3-methoxy-4-phenylmethoxyphenyl)methyl]-6-phenylmethoxyisoquinoline |
| Molecular Weight (g/mol) | 491.58 |
| Formula Weight | 491.2097 |