Learn More
7-Hydroxyindole, 95%
Product Code. 15773467 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15773467 1 g 1g
15783467 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15773467 Supplier Thermo Scientific Acros Supplier No. 462290010
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
CAS: 2380-84-9 | C8H7NO | 133.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2380-84-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key ORVPXPKEZLTMNW-UHFFFAOYSA-N
Synonym 7-hydroxyindole, 7-indolol, 7-hydroxy-1h-indole, indol-7-ol, 7-hydroxy-indole, pubchem7345, 7-hydroxylindole
PubChem CID 2737651
IUPAC Name 1H-indol-7-ol
SMILES C1=CC2=C(C(=C1)O)NC=C2
Specifications
Melting Point 85.0°C to 87.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 133.15
Percent Purity 95%
Chemical Name or Material 7-Hydroxyindole