missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Coumarin-d5, TRC

Product Code. 30397873
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397873 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397873 Supplier Toronto Research Chemicals Supplier No. TRCH92487625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Hydroxy Coumarin-d5
CAS 1215373-23-1
Purity Grade Notes HPLC
Molecular Formula C9HD5O3
Quantity 25 mg
Synonym 7-Hydroxy-2H-1-benzopyran-2-one-d5, 7-Hydroxy-2-chromenone-d5, 7-Oxycoumarin-d5, Hydrangin-d5, Hydrangine-d5, NSC 19790-d5, Skimmetin-d5, Skimmetine-d5, Umbelliferone-d5
InChI Formula InChI=1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H/i1D,2D,3D,4D,5D
SMILES OC1=C([2H])C([2H])=C2C(OC(C([2H])=C2[2H])=O)=C1[2H]
Molecular Weight (g/mol) 167.17
Formula Weight 167.0631
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.