Learn More
7-Ethoxyresorufin N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Ethoxyresorufin N-Oxide |
| CAS | 3705-80-4 |
| Molecular Formula | C14H11NO4 |
| Quantity | 5 mg |
| Synonym | Resazurin Ethyl Ether, 7-Ethoxy-3H-phenoxazin-3-one 10-Oxide |
| InChI Formula | InChI=1S/C14H11NO4/c1-2-18-10-4-6-12-14(8-10)19-13-7-9(16)3-5-11(13)15(12)17/h3-8H,2H2,1H3 |
| SMILES | CCOc1ccc2c(OC3=CC(=O)C=CC3=N2=O)c1 |
| IUPAC Name | 7-ethoxy-10-oxidophenoxazin-10-ium-3-one |
| Molecular Weight (g/mol) | 257.24 |
| Formula Weight | 257.0688 |