Learn More
Afora™ 7/E Round Bottom Flask
Product Code. 12060058
Change view
Click to view available options
Capacity: 250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12060058 Capacity: 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12060058 Supplier Afora™ Supplier No. 7/E
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use the Afora™ Round Bottom Flask for chemical or biochemical work in the laboratory.
  • Weight: 0.123kg
  • Metric Capacity: 250mL
Specifications
Description Weight: 0.123kg
Capacity (Metric) 250 mL
Material Glass
No. per Case 1 Each
Bottom Shape Round