Learn More
7-Chloro-1,3-dihydro-5-(2-fluorophenyl)-2-nitromethylene-2H-1,4-benzodiazepine 4-Oxide, TRC
Product Code. 30010494
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010494 100 mg 100mg
30007387 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30010494 Supplier Toronto Research Chemicals Supplier No. TRCC366465100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60656-76-0
Molecular Formula C16H11ClFN3O3
Molecular Weight (g/mol) 347.73
Synonym 7-Chloro-5-(2-fluorophenyl)-2,3-dihydro-2-(nitromethylene)-1H-1,4-benzodiazepine 4-Oxide, Midazolam Intermediate II
IUPAC Name 7-chloro-1-(3-fluorophenyl)-3-methylidene-2-nitro-4-oxido-2H-1,4-benzodiazepin-4-ium
SMILES C=C1C(N(C2=C(C=C(C=C2)Cl)C=[N+]1[O-])C3=CC(=CC=C3)F)[N+](=O)[O-]
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C16H11ClFN3O3/c1-10-16(21(23)24)20(14-4-2-3-13(18)8-14)15-6-5-12(17)7-11(15)9-19(10)22/h2-9,16H,1H2
Formula Weight 259.016
Chemical Name or Material 7-Chloro-5-(2-fluorophenyl)-2,3-dihydro-2-(nitromethylene)-1H-1,4-benzodiazepine 4-Oxide