Learn More
7-(But-2-yn-1-yl)-3-methyl-8-((4-methylquinazolin-2-yl)methoxy)-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 7-(But-2-yn-1-yl)-3-methyl-8-((4-methylquinazolin-2-yl)methoxy)-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione |
| CAS | 2469626-53-5 |
| Molecular Formula | C30 H26 N8 O3 |
| Quantity | 10 mg |
| Synonym | 7-(But-2-yn-1-yl)-3-methyl-8-((4-methylquinazolin-2-yl)methoxy)-1-((4-methylquinazolin-2-yl)methyl)-3,7-dihydro-1H-purine-2,6-dione, Linagliptin Impurity |
| InChI Formula | InChI=1S/C30H26N8O3/c1-5-6-15-37-26-27(35-29(37)41-17-25-32-19(3)21-12-8-10-14-23(21)34-25)36(4)30(40)38(28(26)39)16-24-31-18(2)20-11-7-9-13-22(20)33-24/h7-14H,15-17H2,1-4H3 |
| SMILES | CC#CCn1c(OCc2nc(C)c3ccccc3n2)nc4N(C)C(=O)N(Cc5nc(C)c6ccccc6n5)C(=O)c14 |
| IUPAC Name | 7-but-2-ynyl-3-methyl-8-[(4-methylquinazolin-2-yl)methoxy]-1-[(4-methylquinazolin-2-yl)methyl]purine-2,6-dione |
| Molecular Weight (g/mol) | 546.579 |
| Formula Weight | 546.213 |