Learn More
7-Benzyloxy Quetiapine, TRC
Product Code. 30336028
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336028 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336028 Supplier Toronto Research Chemicals Supplier No. TRCB2799105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-Benzyloxy Quetiapine
CAS 329217-58-5
Molecular Formula C28H31N3O3S
Quantity 5 mg
Synonym 2-[2-[4-[7-(Phenylmethoxy)dibenzo[b,f][1,4]thiazepin-11-yl]-1-piperazinyl]ethoxy]ethanol
InChI Formula InChI=1S/C28H31N3O3S/c32-17-19-33-18-16-30-12-14-31(15-13-30)28-24-8-4-5-9-26(24)35-27-20-23(10-11-25(27)29-28)34-21-22-6-2-1-3-7-22/h1-11,20,32H,12-19,21H2
SMILES OCCOCCN1CCN(C2=NC3=CC=C(OCC4=CC=CC=C4)C=C3SC5=CC=CC=C25)CC1
IUPAC Name 7-Benzyloxy Quetiapine
Molecular Weight (g/mol) 489.63
Formula Weight 489.21